Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197158.1 | NP_001184087.1 | 2 - 3 | 272 | 86 | |
XM_005615761.3 | XP_005615818.1 | 3 - 4 | 407 | 86 | ||
GenBank mRNA | GQ303578.1 | - | 2 - 3 | 272 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |