Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205169.1 | NP_001192098.1 | 4 - 5 | 528 | 94 | |
GenBank mRNA | DN371994.1 | - | 4 - 5 | 528 | 94 | |
DN749101.1 | - | 3 - 4 | 237 | 94 | ||
EU909456.1 | - | 4 - 5 | 503 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |