Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205169.1 | NP_001192098.1 | 5 - 6 | 669 | 108 | |
GenBank mRNA | CO697123.1 | - | 1 - 2 | 341 | 110 | |
DN749101.1 | - | 4 - 5 | 378 | 108 | ||
EU909456.1 | - | 5 - 6 | 644 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |