Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205190.2 | NP_001192119.1 | 4 - 5 | 777 | 73 | |
XM_005633260.3 | XP_005633317.1 | 5 - 6 | 928 | 73 | ||
XM_022407407.1 | XP_022263115.1 | 6 - 7 | 1085 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |