Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205115.1 | NP_001192044.1 | 4 | 1353 | 114 | |
XM_022412223.1 | XP_022267931.1 | 5 | 1414 | 114 | ||
XM_022412224.1 | XP_022267932.1 | 4 | 1288 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |