Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024636.2 | NP_001019807.1 | 1 | 453 | 66 | |
GenBank mRNA | AB175612.1 | - | 2 - 3 | 247 | 66 | |
AB175614.1 | - | 2 - 3 | 247 | 66 | ||
GR893024.1 | - | 1 | 249 | 66 | ||
GR901969.1 | - | 2 - 3 | 453 | 66 |
Gene Symbol: LOC102155200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634336.2 | XP_005634393.1 | 1 | 452 | 66 | |
GenBank mRNA | AB175613.1 | - | 1 | 247 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Inventoried Both primers and probe map within a single exon |