Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251949.1 | NP_001238878.1 | 5 - 6 | 416 | 66 | |
GenBank mRNA | DN269529.1 | - | 5 - 6 | 401 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |