Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMBIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252399.1 | NP_001239328.1 | 5 - 6 | 409 | 83 | |
XM_005636692.3 | XP_005636749.1 | 5 - 6 | 515 | 83 | ||
XM_022410856.1 | XP_022266564.1 | 5 - 6 | 464 | 83 | ||
GenBank mRNA | DN880529.1 | - | 5 - 6 | 409 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |