Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252410.1 | NP_001239339.1 | 4 - 5 | 352 | 83 | |
GenBank mRNA | DN433890.1 | - | 4 - 5 | 329 | 83 | |
DT536092.1 | - | 4 - 5 | 419 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |