Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254715.1 | NP_001241644.1 | 6 - 7 | 679 | 116 |
Gene Symbol: HAVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417343.1 | XP_022273051.1 | 8 - 9 | 1122 | 116 | |
XM_022417344.1 | XP_022273052.1 | 8 - 9 | 1002 | 116 | ||
XM_022417347.1 | XP_022273055.1 | 7 - 8 | 1062 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |