Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270887.1 | NP_001257816.1 | 5 - 6 | 549 | 93 | |
XM_022423975.1 | XP_022279683.1 | 8 - 9 | 786 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |