Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252417.1 | NP_001239346.1 | 1 | 421 | 90 | |
XM_005617245.3 | XP_005617302.1 | 1 | 592 | 90 | ||
XM_014106713.2 | XP_013962188.1 | 1 | 580 | 90 | ||
GenBank mRNA | DN392560.1 | - | 1 | 421 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |