Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421812.1 | XP_022277520.1 | 2 - 3 | 680 | 120 | |
XM_547729.6 | XP_547729.2 | 1 - 2 | 1417 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Probe spans exons |