Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505560.2 | XP_003505608.1 | 2 - 3 | 1831 | 62 | |
XM_007646056.1 | XP_007644246.1 | 3 - 4 | 1716 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |