Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aagab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500482.2 | XP_003500530.1 | 5 - 6 | 612 | 64 | |
GenBank mRNA | JI870442.1 | - | 5 - 6 | 576 | 64 | |
JP046707.1 | - | 5 - 6 | 576 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |