Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ism1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504337.2 | XP_003504385.1 | 6 - 7 | 867 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Protease 3' Most Recommended Assay Probe spans exons |