Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507694.2 | XP_003507742.1 | 2 - 3 | 472 | 78 | |
GenBank mRNA | JI873741.1 | - | 2 - 3 | 453 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |