Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650817.1 | XP_007649007.1 | 3 - 4 | 493 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Protease Detect Genomic DNA Recommended Assay Probe spans exons |