Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Supt4h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503794.2 | XP_003503842.1 | 3 - 4 | 329 | 78 | |
GenBank mRNA | JI892820.1 | - | 3 - 4 | 313 | 78 | |
JP041585.1 | - | 3 - 4 | 313 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |