Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244058.1 | NP_001230987.1 | 7 - 8 | 870 | 79 | |
XM_007651531.1 | XP_007649721.1 | 7 - 8 | 987 | 79 | ||
GenBank mRNA | DQ202703.1 | - | 7 - 8 | 870 | 79 | |
JI873079.1 | - | 7 - 8 | 969 | 79 | ||
JP061543.1 | - | 7 - 8 | 969 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |