Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246717.1 | NP_001233646.1 | 1 - 2 | 183 | 100 | |
XM_007654978.1 | XP_007653168.1 | 1 - 2 | 263 | 100 | ||
GenBank mRNA | AJ428867.1 | - | 1 - 2 | 183 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |