Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282002.1 | NP_001268931.1 | 5 - 6 | 647 | 73 | |
NM_001282003.1 | NP_001268932.1 | 4 - 5 | 584 | 73 | ||
GenBank mRNA | JI877748.1 | - | 3 - 4 | 492 | 73 | |
JP062696.1 | - | 3 - 4 | 492 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |