Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243990.1 | NP_001230919.1 | 8 - 9 | 1164 | 68 | |
GenBank mRNA | AB042238.1 | - | 8 - 9 | 1164 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |