Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246826.1 | NP_001233755.1 | 4 - 5 | 253 | 83 | |
XM_007647383.1 | XP_007645573.1 | 4 - 5 | 268 | 83 | ||
GenBank mRNA | JI877771.1 | - | 4 - 5 | 270 | 83 | |
JP037986.1 | - | 4 - 5 | 270 | 83 | ||
U31241.1 | - | 4 - 5 | 253 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |