Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246829.1 | NP_001233758.1 | 13 - 14 | 1823 | 101 | |
GenBank mRNA | JI875975.1 | - | 13 - 14 | 1698 | 101 | |
JP065007.1 | - | 13 - 14 | 1698 | 101 | ||
U92313.1 | - | 13 - 14 | 1823 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |