Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244016.1 | NP_001230945.1 | 5 - 6 | 488 | 129 | |
GenBank mRNA | JI884329.1 | - | 5 - 6 | 129 | ||
JP060064.1 | - | 5 - 6 | 129 | |||
K01164.1 | - | 5 - 6 | 488 | 129 | ||
K01165.1 | - | 5 - 6 | 489 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |