Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246708.1 | NP_001233637.1 | 6 - 7 | 1400 | 54 | |
GenBank mRNA | AB016930.1 | - | 6 - 7 | 1400 | 54 | |
JI878307.1 | - | 1 - 2 | 312 | 54 | ||
JP050879.1 | - | 1 - 2 | 312 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |