Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246732.1 | NP_001233661.1 | 2 - 3 | 1056 | 92 | |
GenBank mRNA | DQ384867.1 | - | 2 - 3 | 1056 | 92 | |
JI871851.1 | - | 1 - 2 | 796 | 92 | ||
JP045397.1 | - | 1 - 2 | 796 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |