Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec22b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246746.1 | NP_001233675.1 | 2 - 3 | 266 | 95 | |
GenBank mRNA | JI875226.1 | - | 2 - 3 | 321 | 95 | |
JP060503.1 | - | 2 - 3 | 321 | 95 | ||
U91742.1 | - | 2 - 3 | 266 | 95 |