Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec22b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246746.1 | NP_001233675.1 | 4 - 5 | 573 | 86 | |
GenBank mRNA | JI875226.1 | - | 4 - 5 | 628 | 86 | |
JP060503.1 | - | 4 - 5 | 628 | 86 | ||
U91742.1 | - | 4 - 5 | 573 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |