Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246784.1 | NP_001233713.1 | 4 - 5 | 307 | 104 | |
GenBank mRNA | AY302220.1 | - | 4 - 5 | 307 | 104 | |
JI885704.1 | - | 5 - 6 | 436 | 104 | ||
JP050082.1 | - | 5 - 6 | 435 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |