Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243980.1 | NP_001230909.1 | 1 | 1125 | 78 | |
XM_007644559.1 | XP_007642749.1 | 3 | 2171 | 78 | ||
XM_007644560.1 | XP_007642750.1 | 3 | 2157 | 78 | ||
XM_007644561.1 | XP_007642751.1 | 2 | 2066 | 78 | ||
GenBank mRNA | AF343963.1 | - | 1 | 1125 | 78 | |
AF343964.1 | - | 1 | 1125 | 78 | ||
AF343965.1 | - | 1 | 1125 | 78 | ||
AF510636.1 | - | 1 | 1126 | 78 | ||
AF510637.1 | - | 1 | 1125 | 78 | ||
AF510638.1 | - | 1 | 1126 | 78 | ||
AF510639.1 | - | 1 | 1125 | 78 | ||
AF510640.1 | - | 1 | 1158 | 78 | ||
JI874524.1 | - | 1 | 313 | 78 | ||
JP045923.1 | - | 1 | 311 | 78 | ||
U65791.1 | - | 1 | 1125 | 78 | ||
U65792.1 | - | 1 | 1125 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Inventoried Both primers and probe map within a single exon |