Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243982.1 | NP_001230911.1 | 3 - 4 | 880 | 115 | |
GenBank mRNA | DQ825405.1 | - | 3 - 4 | 880 | 115 | |
JI891225.1 | - | 3 - 4 | 822 | 115 | ||
JP053455.1 | - | 3 - 4 | 822 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |