Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243989.1 | NP_001230918.1 | 9 - 10 | 1124 | 107 | |
XM_007646774.1 | XP_007644964.1 | 10 - 11 | 1288 | 107 | ||
XM_007646775.1 | XP_007644965.1 | 8 - 9 | 1063 | 107 | ||
XM_007646776.1 | XP_007644966.1 | 10 - 11 | 1288 | 107 | ||
XM_007646777.1 | XP_007644967.1 | 10 - 11 | 1287 | 107 | ||
GenBank mRNA | M60041.1 | - | 9 - 10 | 1124 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter 3' Most Recommended Assay Probe spans exons |