Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244042.1 | NP_001230971.1 | 1 - 2 | 150 | 74 | |
GenBank mRNA | JI895124.1 | - | 1 - 2 | 125 | 74 | |
JP063507.1 | - | 1 - 2 | 125 | 74 | ||
L38991.1 | - | 1 - 2 | 150 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |