Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246694.2 | NP_001233623.1 | 3 - 4 | 739 | 73 | |
XM_007643277.1 | XP_007641467.1 | 4 - 5 | 1619 | 73 | ||
GenBank mRNA | AY117536.1 | - | 3 - 4 | 742 | 73 | |
JI896241.1 | - | 2 - 3 | 219 | 73 | ||
JP059857.1 | - | 2 - 3 | 219 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |