Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246736.1 | NP_001233665.1 | 3 - 4 | 354 | 104 | |
GenBank mRNA | JI878699.1 | - | 3 - 4 | 369 | 104 | |
JP046069.1 | - | 3 - 4 | 369 | 104 | ||
U05959.1 | - | 3 - 4 | 354 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |