Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244078.1 | NP_001231007.1 | 2 - 3 | 433 | 105 | |
XM_007640186.1 | XP_007638376.1 | 2 - 3 | 524 | 105 | ||
GenBank mRNA | AF527629.1 | - | 2 - 3 | 433 | 105 | |
JI868716.1 | - | 2 - 3 | 440 | 105 | ||
JP061244.1 | - | 2 - 3 | 440 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |