Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244117.1 | NP_001231046.1 | 4 - 5 | 466 | 113 | |
GenBank mRNA | HQ290320.1 | - | 4 - 5 | 466 | 113 | |
JI864695.1 | - | 3 - 4 | 440 | 113 | ||
JP063222.1 | - | 3 - 4 | 440 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter 3' Most Detect Genomic DNA Probe spans exons |