Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246675.1 | NP_001233604.1 | 3 - 4 | 309 | 80 | |
XM_007622788.1 | XP_007620978.1 | 3 - 4 | 322 | 80 | ||
GenBank mRNA | JI876731.1 | - | 3 - 4 | 271 | 80 | |
JP046177.1 | - | 3 - 4 | 271 | 80 | ||
X98066.1 | - | 3 - 4 | 309 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |