Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246684.1 | NP_001233613.1 | 2 - 3 | 658 | 93 | |
GenBank mRNA | AF113703.1 | - | 2 - 3 | 658 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |