Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246769.1 | NP_001233698.1 | 3 - 4 | 862 | 64 | |
GenBank mRNA | AF306662.2 | - | 3 - 4 | 862 | 64 | |
JI866461.1 | - | 3 - 4 | 579 | 64 | ||
JP048238.1 | - | 3 - 4 | 579 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Detect Genomic DNA Recommended Assay Probe spans exons |