Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244054.1 | NP_001230983.1 | 4 - 5 | 468 | 75 | |
GenBank mRNA | DQ202702.1 | - | 4 - 5 | 468 | 75 | |
JI873415.1 | - | 4 - 5 | 459 | 75 | ||
JP043361.1 | - | 4 - 5 | 459 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |