Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244118.1 | NP_001231047.1 | 2 - 3 | 419 | 65 | |
GenBank mRNA | DQ116786.1 | - | 2 - 3 | 419 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Probe spans exons |