Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246692.1 | NP_001233621.1 | 4 - 5 | 447 | 113 | |
GenBank mRNA | AF351131.1 | - | 4 - 5 | 447 | 113 | |
JI891533.1 | - | 3 - 4 | 357 | 113 | ||
JP056719.1 | - | 3 - 4 | 356 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |