Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fshb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246712.1 | NP_001233641.1 | 2 - 3 | 219 | 61 | |
GenBank mRNA | AB248599.1 | - | 2 - 3 | 219 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |