Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252299.1 | NP_001239228.1 | 5 - 6 | 508 | 76 | |
GenBank mRNA | JF935144.1 | - | 5 - 6 | 508 | 76 | |
JI862432.1 | - | 5 - 6 | 585 | 76 | ||
JP041261.1 | - | 5 - 6 | 585 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |