Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501088.2 | XP_003501136.1 | 3 - 4 | 844 | 63 | |
GenBank mRNA | JI882839.1 | - | 3 - 4 | 823 | 63 | |
JP041523.1 | - | 3 - 4 | 823 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |