Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasl11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514140.2 | XP_003514188.1 | 1 - 2 | 215 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |