Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam83d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497113.2 | XP_003497161.1 | 3 - 4 | 797 | 73 | |
GenBank mRNA | JI891180.1 | - | 2 - 3 | 157 | 73 | |
JP039991.1 | - | 2 - 3 | 157 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |